Anti-LUM Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60212-100UL A60212-50UL
77716-139EA 529.22 CAD
77716-139 77716-140
Anti-LUM Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: LUM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR