Anti-BMP6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60186-100UL A60186-50UL
77716-100EA 529.22 CAD
77716-100 77716-101
Anti-BMP6 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: BMP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR