Anti-SEMA4C Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60165-100UL A60165-50UL
77716-063EA 529.22 CAD
77716-063 77716-064
Anti-SEMA4C Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: SEMA4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
New Product
Order Now

SPECIFICATIONS

Antigen symbol SEMA4C
Environmentally Preferable
Antigen synonyms KIAA1739|SEMAF|SEMA4C|SEMACL1|Sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C|Semaphorin-4C|UNQ5855/PRO34487|SEMAI|S4c|M-SEMA-F|SEM4C_HUMAN
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Gene ID Q9C0C4
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Immunogen Fusion protein of Human SEMA4C
Form Liquid
Purification Antigen affinity purified
Storage buffer PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR