Anti-DSC2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60143-100UL A60143-50UL
77716-026EA 529.22 CAD
77716-026 77716-027
Anti-DSC2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: DSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR