Anti-PIK3C2G Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60083-100UG A60083-50UG
77705-610EA 400.93 CAD
77705-610 77715-927
Anti-PIK3C2G Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, ELISA:1:10000

Type:
Antigen: PIK3C2G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR