Anti-PRC1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60055-100UG A60055-50UG
77705-608EA 400.93 CAD
77705-608 77715-880
Anti-PRC1 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-2000, ELISA:1:10000-20000

Type:
Antigen: PRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR