Anti-ATOX1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60037-100UL A60037-50UL
77715-851EA 529.22 CAD
77715-851 77715-852
Anti-ATOX1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: ATOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR