Anti-ARHGAP4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60032-100UL A60032-50UL
77715-842EA 529.22 CAD
77715-842 77715-843
Anti-ARHGAP4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: ARHGAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR