Anti-CDKN1A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49106-50UL A49106-100UL
77715-815EA 360.84 CAD
77715-815 77715-814
Anti-CDKN1A Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: CDKN1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR