Anti-FEM1A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49101-50UL A49101-100UL
77715-807EA 360.84 CAD
77715-807 77715-806
Anti-FEM1A Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: FEM1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR