Anti-PLXNA3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49089-100UL A49089-50UL
77715-786EA 529.22 CAD
77715-786 77715-787
Anti-PLXNA3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: PLXNA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR