Anti-TERT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49058-100UL A49058-50UL
77715-737EA 529.22 CAD
77715-737 77715-738
Anti-TERT Rabbit Polyclonal Antibody
Antibodies
ELISA:1:3000-1:10000, IHC:1:50-1:200

Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR