Anti-PLAT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49026-50UL A49026-100UL
77715-684EA 360.84 CAD
77715-684 77715-683
Anti-PLAT Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: PLAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR