Anti-RASGEF1A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49025-50UL A49025-100UL
77715-682EA 360.84 CAD
77715-682 77715-681
Anti-RASGEF1A Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:10-1:50

Type:
Antigen: RASGEF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR