Anti-MC4R Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48993-50UL A48993-100UL
77715-627EA 360.84 CAD
77715-627 77715-626
Anti-MC4R Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:50-1:200

Type:
Antigen: MC4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR