Anti-SCN10A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48933-50UL A48933-100UL
77715-526EA 360.84 CAD
77715-526 77715-525
Anti-SCN10A Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:25-1:100

Type:
Antigen: SCN10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR