Anti-CXCR4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48914-50UL A48914-100UL
77715-494EA 360.84 CAD
77715-494 77715-493
Anti-CXCR4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
New Product
Order Now

SPECIFICATIONS

Antigen symbol CXCR4
Environmentally Preferable
Antigen synonyms HM89|SDF-1 receptor|C-X-C chemokine receptor type 4|Leukocyte-derived seven transmembrane domain receptor|WHIM|NPYR|CXCR4_HUMAN|Probable G protein coupled receptor lcr1 homolog|NPYRL|SEVEN-TRANSMEMBRANE-SEGMENT RECEPTOR|Chemokine (C X C motif) receptor 4|CXCR-4|Neuropeptide Y receptor Y3|CXCR4|Stromal cell derived factor 1 receptor|CXC-R4|D2S201E|NPY3R|NPYY3R|CD184 antigen|LCR1|Fusin|HSY3RR|Chemokine CXC Motif Receptor 4|SDF 1 receptor|LAP 3|Leukocyte derived seven transmembrane domain receptor|CD184|Lipopolysaccharide associated protein 3|WHIMS|LAP3|LESTR|FB22|NPYY3|Stromal cell-derived factor 1 receptor
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID P61073
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide of Human CXCR4
Form Liquid
Purification Antigen affinity purified
Storage buffer PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR