Anti-VCAN Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48889-100UG A48889-50UG
77705-378EA 400.93 CAD
77705-378 77715-453
Anti-VCAN Rabbit Polyclonal Antibody
Antibodies
IHC:1:50-300, ELISA:1:10000-20000

Type:
Antigen: VCAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR