Anti-MAP3K5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48829-100UL A48829-50UL
77715-350EA 529.22 CAD
77715-350 77715-351
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:25-1:100

Type:
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR