Anti-CIDEC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48751-50UL A48751-100UL
77715-218EA 360.84 CAD
77715-218 77715-217
Anti-CIDEC Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: CIDEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR