Anti-ACTB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48743-100UL A48743-50UL
77715-203EA 529.22 CAD
77715-203 77715-204
Anti-ACTB Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:500-1:2000

Type:
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR