Anti-CD33 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48690-50UL A48690-100UL
77715-111EA 360.84 CAD
77715-111 77715-110
Anti-CD33 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: CD33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR