Anti-PLS3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48635-100UL A48635-50UL
77715-017EA 529.22 CAD
77715-017 77715-018
Anti-PLS3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: PLS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR