Anti-ITPR1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48628-100UL A48628-50UL
77715-004EA 529.22 CAD
77715-004 77715-005
Anti-ITPR1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:50-1:200

Type:
Antigen: ITPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR