Anti-HNF4A/HNF4G (acetyl-Lys127/79) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48623-100UG A48623-50UG
77705-369EA 400.93 CAD
77705-369 77714-996
Anti-HNF4A/HNF4G (acetyl-Lys127/79) Rabbit Polyclonal Antibody
Antibodies
WB:1:500-2000, ELISA:1:10000-20000

Type:
Antigen: HNF4A/HNF4G (acetyl-Lys127/79)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR