Anti-ADA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48611-50UL A48611-100UL
77714-978EA 360.84 CAD
77714-978 77714-977
Anti-ADA Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:15-1:50

Type:
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR