Anti-REG3A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48605-100UL A48605-50UL
77714-966EA 529.22 CAD
77714-966 77714-967
Anti-REG3A Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: REG3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR