Anti-DNAJC19 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A65383-100UL A65383-50UL
77690-467EA 529.22 CAD
77690-467 77725-815
Anti-DNAJC19 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: DNAJC19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR