Anti-CHMP4C Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A65369-100UG A65369-50UG
77725-789EA 529.22 CAD
77725-789 77725-790
Anti-CHMP4C Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CHMP4C
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR