Anti-CELSR3 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A65246-50UL A65246-100UL
77725-550EA 360.84 CAD
77725-550 77725-549
Anti-CELSR3 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CELSR3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR