Anti-CELSR3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A65243-50UL A65243-100UL
77725-544EA 360.84 CAD
77725-544 77725-543
Anti-CELSR3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:200-1:500

Type:
Antigen: CELSR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR