Anti-PPIL4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A65196-100UG A65196-50UG
77690-455EA 529.22 CAD
77690-455 77725-453
Anti-PPIL4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200

Type:
Antigen: PPIL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR