Anti-ACMSD Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A65170-100UG A65170-50UG
77725-407EA 529.22 CAD
77725-407 77725-408
Anti-ACMSD Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ACMSD
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR