Anti-ACMSD Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A65169-100UG A65169-50UG
77690-449EA 529.22 CAD
77690-449 77725-406
Anti-ACMSD Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: ACMSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR