Anti-ATAD1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A65094-100UG A65094-50UG
77725-266EA 529.22 CAD
77725-266 77725-267
Anti-ATAD1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ATAD1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR