Anti-BEND7 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A65053-100UG A65053-50UG
77725-186EA 529.22 CAD
77725-186 77725-187
Anti-BEND7 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: BEND7
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR