Anti-NGEF Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A65039-50UL A65039-100UL
77725-161EA 360.84 CAD
77725-161 77725-160
Anti-NGEF Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NGEF
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR