Anti-RPRD1A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64937-100UG A64937-50UG
77690-420EA 529.22 CAD
77690-420 77724-971
Anti-RPRD1A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: RPRD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR