Anti-ALG14 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64850-100UG A64850-50UG
77724-804EA 529.22 CAD
77724-804 77724-805
Anti-ALG14 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: ALG14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR