Anti-Cbs Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A64732-50UG A64732-100UG
77724-576EA 360.84 CAD
77724-576 77724-575
Anti-Cbs Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: Cbs
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR