Anti-Cbs Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A64731-100UG A64731-50UG
77724-573EA 529.22 CAD
77724-573 77724-574
Anti-Cbs Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Cbs
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR