Anti-Cbs Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64729-100UG A64729-50UG
77690-405EA 529.22 CAD
77690-405 77724-570
Anti-Cbs Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF:1:50-1:200

Type:
Antigen: Cbs
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR