Anti-CTNNBL1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64722-100UL A64722-50UL
77690-403EA 529.22 CAD
77690-403 77724-558
Anti-CTNNBL1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: CTNNBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR