Anti-NEU4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64696-50UG A64696-100UG
77724-509EA 360.84 CAD
77724-509 77724-508
Anti-NEU4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:200-1:500, IF:1:50-1:200

Type:
Antigen: NEU4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR