Anti-TXNDC5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64545-100UL A64545-50UL
77690-380EA 529.22 CAD
77690-380 77724-229
Anti-TXNDC5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200, IP:1:200-1:2000

Type:
Antigen: TXNDC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR