Anti-POGLUT1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A61466-50UL A61466-100UL
77718-451EA 360.84 CAD
77718-451 77718-450
Anti-POGLUT1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: POGLUT1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR