Anti-P3H2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61170-100UG A61170-50UG
77690-077EA 529.22 CAD
77690-077 77717-902
Anti-P3H2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:2000-1:5000, IHC:1:20-1:200

Type:
Antigen: P3H2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR