Anti-RBM45 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61084-100UG A61084-50UG
77690-069EA 529.22 CAD
77690-069 77717-739
Anti-RBM45 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200

Type:
Antigen: RBM45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR