Anti-CMTR2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61047-50UG A61047-100UG
77717-670EA 360.84 CAD
77717-670 77717-669
Anti-CMTR2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: CMTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR