Anti-CCDC103 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61014-100UG A61014-50UG
77690-062EA 529.22 CAD
77690-062 77717-606
Anti-CCDC103 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: CCDC103
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR